Masterflex® Correlated Variable Area Flowmeters, PTFE Housing, 65-mm Scale
Designed for high-purity applications with flexibility for a broad array of gases
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
The substitution of metal fittings with PTFE eliminates a potential contamination source for high-purity applications. The glass tube utilizes a fused ceramic scale for a precise, permanent measuring guide. In addition, a vertical-tangential locator line provides readings with hairline accuracy. Further improving readability is a front shield that magnifies the scale 16%. An anodized aluminum frame protects the heavy-walled glass flowtube; a white acrylic backplate protects and enhances viewing in the protective frame.
Select a flowmeter with a valve for flow rate control. The standard valve is suitable where high resolution metering is not essential. The high precision valve features a fine-adjust 16-turn “non-rising stem” to more accurately set your desired flow rate.
All flowmeters come with correlation data sheets for water and numerous gases (listed below) at standard temperature and pressure (STP).
$249.60 - $360.90USD / Each
35 variations of this product are available.
See All Product Options Below
35 PRODUCT OPTIONS
Compareitem specifications
Material Fitting Process Connection (NPT(F)) Description
Availability Pricing
COMPARE
PTFE and Kel-F® 0 65 mm Correlated PTFE Flowmeter, Valve, Sapphire, 042-07
$305.00USD / Each
COMPARE
PTFE and Kel-F® 0 65 mm Correlated PTFE Flowmeter, Valved, Sapphire, 032-15
$305.00USD / Each
COMPARE
PTFE and Kel-F® 0 65 mm Correlated PTFE Flowmeter, Valved, Glass, 012-10
$305.00USD / Each
COMPARE
PTFE and Kel-F® 0 65 mm Correlated PTFE Flowmeter, Valved, Sapphire, 012-10
$305.00USD / Each
COMPARE
PTFE and Kel-F® 0 65 mm Correlated PTFE Flowmeter, Valved, Glass, 052-01
$305.00USD / Each
COMPARE
PTFE and Kel-F® 0 65 mm Correlated PTFE Flowmeter, Valved, Sapphire, 052-01
$305.00USD / Each
COMPARE
PTFE and Kel-F® 0 65 mm Correlated PTFE Flowmeter, Valved, Glass, 023-92
$295.00USD / Each
COMPARE
PTFE and Kel-F® 0 65 mm Correlated PTFE Flowmeter, Valved, Glass, 013-88
$305.00USD / Each
COMPARE
PTFE and Kel-F® 0 65 mm Correlated PTFE Flowmeter, Valved, Sapphire, 013-88
$310.00USD / Each
COMPARE
PTFE and Kel-F® 0 65 mm Correlated PTFE Flowmeter, Valved, Sapphire, 014-96
$315.00USD / Each
COMPARE
PTFE and Kel-F® 0 65 mm Correlated PTFE Flowmeter, No Valve, Sapphire 032-15
$249.60USD / Each
COMPARE
PTFE and Kel-F® 0 65 mm Correlated PTFE Flowmeter, No Valve, Sapphire 012-10
$249.60USD / Each
COMPARE
PTFE and Kel-F® 0 65 mm Correlated PTFE Flowmeter, No valve, 61 mL/min water flow rate
$262.60USD / Each
COMPARE
PTFE and Kel-F® 0 65 mm Correlated PTFE Flowmeter, HR Valve, Glass 022-13
$344.80USD / Each
COMPARE
PTFE and Kel-F® 0 65 mm Correlated PTFE Flowmeter, HR Valve, Glass 012-10
$344.80USD / Each
COMPARE
PTFE and Kel-F® 0 65 mm Correlated PTFE Flowmeter, HR Valve, Sapphire 012-10
$344.80USD / Each
COMPARE
PTFE and Kel-F® 0 65 mm Correlated PTFE Flowmeter, HR Valve, Glass 052-01
$344.80USD / Each
COMPARE
PTFE and Kel-F® 0 65 mm Correlated PTFE Flowmeter, HR Valve, Sapphire 052-01
$344.80USD / Each
COMPARE
PTFE and Kel-F® 0 65 mm Correlated PTFE Flowmeter, HR Valve, Glass 014-96
$344.80USD / Each
COMPARE
PTFE and Kel-F® 0 65 mm Correlated PTFE Flowmeter, HR Valve, Sapphire 064-63
$360.90USD / Each
Showing 1 - 20 of 35