Masterflex® Correlated Variable-Area Flowmeters with Valve, 65-mm Scale
Read more easily with 16% magnification of scale
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Each flowmeter consists of a heavy-walled glass flowtube mounted in a frame with white acrylic back plate (1/8" thick). Aluminum Flowmeters are economical and good for general use with noncorrosive gases and liquids. Brass Flowmeters are economical and good for use with water. 316 Stainless Steel Flowmeters feature excellent chemical compatibility.
Select a flowmeter with a high-resolution valve for superior flow rate control—ideal for low-flow applications or for any application where you need precise flow control. All flowmeters come with correlation data sheets for water and air at standard temperature and pressure (STP). We can also supply calibration data for oxygen, nitrogen, hydrogen, helium, CO2, argon, other liquids and gases, and for conditions other than STP — call our Application Specialists for more information.
$137.40 - $270.80USD / Each
57 variations of this product are available.
See All Product Options Below
57 PRODUCT OPTIONS
Compareitem specifications
Material Housing Max Flow Rate (mL/min) Process Connection (NPT(F))
Availability Pricing
COMPARE
Aluminum 48.7 0
$137.40USD / Each
COMPARE
Aluminum 299 0
$138.00USD / Each
COMPARE
Aluminum 1249 0
$138.00USD / Each
COMPARE
Aluminum 2678 0
$138.00USD / Each
COMPARE
Aluminum 4922 0
$138.00USD / Each
COMPARE
Aluminum 12058 0
$138.00USD / Each
COMPARE
Aluminum 13153 0
$138.00USD / Each
COMPARE
316 SS 145 0
$270.80USD / Each
COMPARE
316 SS 299 0
$270.80USD / Each
COMPARE
316 SS 202.1 0
$270.80USD / Each
COMPARE
316 SS 522 0
$270.80USD / Each
COMPARE
316 SS 992 0
$270.80USD / Each
COMPARE
316 SS 2040 0
$270.80USD / Each
COMPARE
316 SS 3990 0
$270.80USD / Each
COMPARE
316 SS 2678 0
$270.80USD / Each
COMPARE
316 SS 6318 0
$270.80USD / Each
COMPARE
316 SS 13153 0
$270.80USD / Each
COMPARE
Brass 2520 0
$179.50USD / Each
COMPARE
Brass 2040 0
$179.50USD / Each
COMPARE
Brass 24680 0
$179.50USD / Each
Showing 1 - 20 of 57