Masterflex® Correlated Variable-Area Flowmeters with Valves, High-Flow, 150-mm
Don’t sacrifice suitability for precision
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. Valved meters have valves mounted at the bottom/inlet, typically used for positive pressure with liquids and gases. The valve can be reconfigured to mount at the top/outlet, often for vacuum applications. All correlated flowmeters come with data sheets for water and air at standard temperature and pressure (STP).
$212.00 - $401.00USD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Material Fitting Process Connection (NPT(F)) Description
Availability Pricing
COMPARE
Aluminum 0 150-mm Correlated Flowmeter, w/ valve, aluminum, 23.742 LPM air
$212.00USD / Each
COMPARE
Aluminum 0 150-mm Correlated Flowmeter, w/ valve, aluminum, 45.227 LPM air
$212.00USD / Each
COMPARE
Aluminum 0 150-mm Correlated Flowmeter, w/ valve, aluminum, 66.346 LPM air
$212.00USD / Each
COMPARE
PTFE and Kel-F® 0 150-mm Correlated Flowmeter, w/ valve, PTFE, 22.536 LPM air
$401.00USD / Each
COMPARE
PTFE and Kel-F® 0 150-mm Correlated Flowmeter, w/ valve, PTFE, 29.56 LPM air
$399.90USD / Each
COMPARE
316 SS 0 150-mm Correlated Flowmeter, w/ valve, 316SS, 23.742 LPM air
$341.80USD / Each
COMPARE
316 SS 0 150-mm Correlated Flowmeter, w/ valve, 316SS, 45.227 LPM air
$343.60USD / Each
COMPARE
316 SS 0 150-mm Correlated Flowmeter, w/ valve, 316SS, 66.346 LPM air
$343.60USD / Each
COMPARE
Chrome-plated brass 0 150-mm Correlated Flowmeter, w/ valve, brass, 23.742 LPM air
$231.60USD / Each
COMPARE
Chrome-plated brass 0 150-mm Correlated Flowmeter, w/ valve, brass, 45.227 LPM air
$231.60USD / Each
COMPARE
Chrome-plated brass 0 150-mm Correlated Flowmeter, w/ valve, brass, 66.346 LPM air
$231.60USD / Each
Showing 1- 11 of 11