Masterflex® Direct English-Reading, Variable-Area Flowmeters for Air/Water, Acrylic Housing, 2", 4", or 5" Scale
Durable single-piece body withstands severe shock and vibration
  • Ideal for process plant applications on air sampling equipment, gas analyzers and chemical feed systems for water treatment
  • Flexible design allows for panel or bench mounting
  • Variable Area with Direct English-Reading Scales
Machined from solid acrylic blocks, these meters have integral metering tubes that provide precise readings even in aggressive plant environments. The meters’ inlet/outlet ports and mounting studs are extended for easy panel installation. An alternate option is a tripod base (sold separately below) which allows for mobility from bench to bench.
Note: There are many additional types of acrylic flowmeters. Contact our Application Specialists for quotes on acrylic meters with special requirements.
$86.50 - $268.00USD / Each
28 variations of this product are available.
See All Product Options Below
28 PRODUCT OPTIONS
Compareitem specifications
Display Type Fluid Type Max Flow Rate (GPH)
Availability Pricing
COMPARE
2" Scale Water 2
$89.00USD / Each
COMPARE
2" Scale Water 10
$86.50USD / Each
COMPARE
2" Scale Water 40
$86.50USD / Each
COMPARE
2" Scale Air 14.961
$86.50USD / Each
COMPARE
2" Scale Air 37.403
$90.00USD / Each
COMPARE
2" Scale Air 1496.1
$86.50USD / Each
COMPARE
4" Scale Water 10
$112.00USD / Each
COMPARE
4" Scale Water 25
$109.00USD / Each
COMPARE
4" Scale Water 60
$109.00USD / Each
COMPARE
4" Scale Water 150
$170.00USD / Each
COMPARE
4" Scale Water 300
$162.00USD / Each
COMPARE
4" Scale Air 37.403
$129.00USD / Each
COMPARE
4" Scale Air 74.805
$105.00USD / Each
COMPARE
4" Scale Air 748.05
$112.00USD / Each
COMPARE
4" Scale Air 1496.1
$113.00USD / Each
COMPARE
4" Scale Air 2244.2
$169.00USD / Each
COMPARE
4" Scale Air 4488.3
$170.00USD / Each
COMPARE
5" Scale Water 1500
$256.00USD / Each
COMPARE
5" Scale Air 11221
$268.00USD / Each
COMPARE
5" Scale Air 44883
$261.00USD / Each
Showing 1 - 20 of 28