Masterflex® Direct Lift 2- and 3-way Solenoid Valves
Ideal for use with high-purity or corrosive fluids
  • All wetted parts made of inert PTFE
  • Minimal dead volume reduces problems with trapped liquids
Rated for 26" Hg vacuum. Maximum fluid temperature is 158°F (70°C). All valves have 15" lead wires. Valves are compact and lightweight — mount them almost anywhere. Valves with a 1/16" orifice have mounting holes at the bottom only and valves with a 3/32" and 5/32" orifice have mounting holes at the top and bottom.
$184.00 - $502.00USD / Each
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
Compareitem specifications
Flow Pattern Inlet Port Power (VDC)
Availability Pricing
COMPARE
Two-Way, Normally Closed 1/4"-28 UNF 12
$184.00USD / Each
COMPARE
Two-Way, Normally Closed 1/4"-28 UNF 24
$185.00USD / Each
COMPARE
Two-Way, Normally Closed 1/4"-28 UNF 12
$230.00USD / Each
COMPARE
Two-Way, Normally Closed 1/4"-28 UNF 24
$230.00USD / Each
COMPARE
Two-Way, Normally Closed 1/8" NPT(F) 12
$448.00USD / Each
COMPARE
Two-Way, Normally Closed 1/8" NPT(F) 24
$448.00USD / Each
COMPARE
Two-Way, Normally Open 1/4"-28 UNF 12
$208.00USD / Each
COMPARE
Two-Way, Normally Open 1/4"-28 UNF 24
$208.00USD / Each
COMPARE
Three-Way 1/4"-28 UNF 12
$205.00USD / Each
COMPARE
Three-Way 1/4"-28 UNF 24
$205.00USD / Each
COMPARE
Three-Way 1/4"-28 UNF 12
$290.00USD / Each
COMPARE
Three-Way 1/4"-28 UNF 24
$290.00USD / Each
COMPARE
Three-Way 1/8" NPT(F) 12
$499.00USD / Each
COMPARE
Three-Way 1/8" NPT(F) 24
$502.00USD / Each
Showing 1- 14 of 14
Need Help with this product?
Call Us At1-847-549-7600
Live chat