Masterflex® Direct Metric-Reading, Variable-Area Flowmeters for Air/Water, Acrylic Housing, 50, 100, or 127 mm Scale
Durable single-piece body withstands severe shock and vibration
  • Ideal for process plant applications on air sampling equipment, gas analyzers and chemical feed systems for water treatment
  • Flexible design allows for panel or bench mounting
  • Variable Area with Direct Metric-Reading Scales
Machined from solid acrylic blocks, these meters have integral metering tubes that provide precise readings even in aggressive plant environments. The meters’ inlet/outlet ports and mounting studs are extended for easy panel installation. An alternate option is a tripod base (sold separately below) which allows for mobility from bench to bench.
Note: There are many additional types of acrylic flowmeters not listed here. Contact our Application Specialists for quotes on acrylic meters with special requirements.
$83.00 - $268.00USD / Each
33 variations of this product are available.
See All Product Options Below
33 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fluid Type Max Flow Rate (mL/min) Process Connection (NPT(F))
Availability Pricing
COMPARE
air 5000 0
$89.00USD / Each
COMPARE
air 25000 0
$86.50USD / Each
COMPARE
air 100000 0
$89.00USD / Each
COMPARE
water 225 0
$109.00USD / Each
COMPARE
water 1500 0
$112.00USD / Each
COMPARE
water 9000
$163.00USD / Each
COMPARE
water 20000
$167.00USD / Each
COMPARE
air 5000 0
$113.00USD / Each
COMPARE
air 10000 0
$116.00USD / Each
COMPARE
air 30000 0
$131.00USD / Each
COMPARE
air 50000 0
$114.00USD / Each
COMPARE
air 100000 0
$115.00USD / Each
COMPARE
air 140000 0
$170.00USD / Each
COMPARE
air 280000 0
$173.00USD / Each
COMPARE
air 560000
$166.00USD / Each
COMPARE
water 15000
$268.00USD / Each
COMPARE
water 36000
$248.00USD / Each
COMPARE
water 75000
$256.00USD / Each
COMPARE
air 700000
$260.00USD / Each
COMPARE
air 1400000
$260.00USD / Each
Showing 1 - 20 of 33