Masterflex® Direct-Reading Variable-Area Flowmeters for Helium, Acrylic
Designed for labs and plant processes where multiple gases are used
  • Easy to read English or metric scales
  • Interchangeable scales for routine gases
  • Customer convertible panel to partial or full in-line mounted configurations
  • Panel mounting by means of hex nuts
  • Easy disassembly and assembly for cleaning
Versatile acrylic flowmeters are designed specifically for helium. Flowmeters are offered with or without built-in low hysteresis needle valves. Panel mount flowmeters without valves or use in partial or full in-line mounted configurations. Flowmeters feature a stable, easy-to-read float for quick measurement readings.
$58.50 - $84.65USD / Each
25 variations of this product are available.
See All Product Options Below
25 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Fitting Max Flow Rate (LPM) Valve
Availability Pricing
COMPARE
Brass 2 No
$58.50USD / Each
COMPARE
Brass 5.5 No
$58.50USD / Each
COMPARE
Brass 8 No
$58.50USD / Each
COMPARE
Brass 55 No
$58.50USD / Each
COMPARE
Brass 110 No
$58.50USD / Each
COMPARE
316 SS 2 No
$64.50USD / Each
COMPARE
316 SS 5.5 No
$64.50USD / Each
COMPARE
316 SS 8 No
$64.50USD / Each
COMPARE
316 SS 110 No
$64.50USD / Each
COMPARE
Brass 5.5 Yes
$67.50USD / Each
COMPARE
Brass 8 Yes
$67.50USD / Each
COMPARE
Brass 18 Yes
$67.50USD / Each
COMPARE
Brass 35 Yes
$67.50USD / Each
COMPARE
Brass 55 Yes
$67.50USD / Each
COMPARE
Brass 110 Yes
$67.50USD / Each
COMPARE
316 SS 2 Yes
$84.65USD / Each
COMPARE
316 SS 5.5 Yes
$84.65USD / Each
COMPARE
316 SS 8 Yes
$84.65USD / Each
COMPARE
316 SS 18 Yes
$84.65USD / Each
COMPARE
316 SS 35 Yes
$84.65USD / Each
Showing 1 - 20 of 25