Masterflex® Direct-Reading Variable-Area Flowmeters for Multiple Gases
Rotating flowtube allows one meter to be used for five gases
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Constructed with durable polycarbonate frame for rugged applications
  • Designed with graduations in English and metric for additional flexibility
  • Use with multiple routine gases for laboratory and industrial applications
Designed for medium flow range applications, these flowmeters incorporate precision-machined glass and brass or stainless steel fittings to provide accurate and economical flow metering solutions. Simply rotate glass flowtube to read the gas you will be using—oxygen, nitrogen, helium, carbon dioxide, and argon.
$381.90 - $662.50USD / Each
80 variations of this product are available.
See All Product Options Below
80 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (LPM) Max Flow Rate (SCFM) Description
Availability Pricing
COMPARE
14 Flowmeter, In-line, With Brass Fittings; 14 SCFM (O2)
$381.90USD / Each
COMPARE
14 Flowmeter, In-line, With Stainless Steel Fittings; 14 SCFM (O2)
$558.25USD / Each
COMPARE
9 Flowmeter, In-line With Valve, With Brass Fittings; 9 SCFM (O2)
$441.00USD / Each
COMPARE
14 Flowmeter, In-line With Valve, With Brass Fittings; 14 SCFM (O2)
$441.00USD / Each
COMPARE
4.5 Multigas Flowmeter, In-Line w/ Valve, SS, 4.5 scfm (O2)
$650.50USD / Each
COMPARE
14 Multigas Flowmeter, In-Line w/ Valve, SS, 14 scfm (O2)
$650.50USD / Each
COMPARE
130 Flowmeter, In-line, With Brass Fittings; 130 SLPM (O2)
$381.90USD / Each
COMPARE
260 Flowmeter, In-line, With Brass Fittings; 260 SLPM (O2)
$381.90USD / Each
COMPARE
400 Flowmeter, In-line, With Brass Fittings; 400 SLPM (O2)
$381.90USD / Each
COMPARE
525 Flowmeter, In-line, With Brass Fittings; 525 SLPM (O2)
$381.90USD / Each
COMPARE
525 Flowmeter, In-line, With Stainless Steel Fittings; 525 SLPM (O2)
$558.25USD / Each
COMPARE
130 Flowmeter, In-line With Valve, With Brass Fittings; 130 SLPM (O2)
$441.00USD / Each
COMPARE
4.5 Flowmeter, Panel-mount, With Brass Fittings; 4.5 SCFM (O2)
$381.90USD / Each
COMPARE
28 Flowmeter, Panel-mount, With Brass Fittings; 28 SCFM (O2)
$381.90USD / Each
COMPARE
18 Multigas Flowmeter, Panel-Mount w/ Valve, SS, 18 scfm (O2)
$662.50USD / Each
COMPARE
400 Flowmeter, Panel-mount, With Brass Fittings; 400 SLPM (O2)
$381.90USD / Each
COMPARE
130 Flowmeter, Panel-mount, With Stainless Steel Fittings; 130 SLPM (O2)
$558.25USD / Each
COMPARE
400 Flowmeter, Panel-mount, With Stainless Steel Fittings; 400 SLPM (O2)
$558.25USD / Each
COMPARE
525 Flowmeter, Panel-mount With Valve, With Brass Fittings; 525 SLPM (O2)
$448.00USD / Each
COMPARE
800 Flowmeter, Panel-mount With Valve, With Brass Fittings; 800 SLPM (O2)
$448.00USD / Each
Showing 1 - 20 of 80