Masterflex® Direct-Reading Variable-Area Flowmeters for Nitrogen, Acrylic
Designed for labs and plant processes where multiple gases are used
  • Easy to read English or metric scales
  • Interchangeable scales for routine gases
  • Customer convertible panel to partial or full in-line mounted configurations
  • Panel mounting by means of hex nuts
  • Easy disassembly and assembly for cleaning
Versatile acrylic flowmeters are designed specifically for nitrogen. Flowmeters are offered with or without built-in low hysteresis needle valves. Panel mount flowmeters without valves or use in partial or full in-line mounted configurations. Flowmeters feature a stable, easy-to-read float for quick measurement readings.
$58.50 - $84.65USD / Each
28 variations of this product are available.
See All Product Options Below
28 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Fitting Max Flow Rate (LPM) Valve
Availability Pricing
COMPARE
Brass 2.75 No
$58.50USD / Each
COMPARE
Brass 3.5 No
$58.50USD / Each
COMPARE
Brass 8 No
$58.50USD / Each
COMPARE
Brass 45 No
$58.50USD / Each
COMPARE
316 SS 3.5 No
$64.50USD / Each
COMPARE
316 SS 8 No
$64.50USD / Each
COMPARE
316 SS 16 No
$64.50USD / Each
COMPARE
316 SS 22 No
$64.50USD / Each
COMPARE
316 SS 45 No
$64.50USD / Each
COMPARE
Brass 1.3 Yes
$67.50USD / Each
COMPARE
Brass 3.5 Yes
$67.50USD / Each
COMPARE
Brass 8 Yes
$67.50USD / Each
COMPARE
Brass 16 Yes
$67.50USD / Each
COMPARE
Brass 22 Yes
$67.50USD / Each
COMPARE
Brass 45 Yes
$67.50USD / Each
COMPARE
316 SS 1.3 Yes
$84.65USD / Each
COMPARE
316 SS 3.5 Yes
$84.65USD / Each
COMPARE
316 SS 8 Yes
$84.65USD / Each
COMPARE
316 SS 22 Yes
$84.65USD / Each
COMPARE
316 SS 45 Yes
$84.65USD / Each
Showing 1 - 20 of 28