Masterflex® Direct-Reading Variable Area Flowmeters for Water, Brass Housing
Don’t sacrifice suitability for precision
  • Direct-reading flowmeters feature a calibrated scale for a specific medium - no need to convert readings to get flow rate│Use with water for laboratory and industrial applications
Use for high pressure industrial applications—install in any configuration, even next to a bend in your pipeline. A calibrated retention spring eliminates the need to mount flowmeters vertically or run straight lengths of pipe prior to or after the meter. These rugged flowmeters are relatively insensitive to shock and vibrations—the spring, piston assembly, and internal magnet compensate for rough process environments.
$267.40 - $799.25USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (mL/min) Min Flow Rate (mL/min) Pipe Size
Availability Pricing
COMPARE
$267.40USD / Each
COMPARE
$267.40USD / Each
COMPARE
$334.60USD / Each
COMPARE
$334.60USD / Each
COMPARE
$509.50USD / Each
COMPARE
$496.50USD / Each
COMPARE
$501.25USD / Each
COMPARE
$799.25USD / Each
COMPARE
$799.25USD / Each
COMPARE
$799.25USD / Each
Showing 1- 10 of 10