Masterflex® Direct-Reading Variable Area Flowmeters for Water, Dual-Scale, 303 SS Housing
Install in any position
  • Direct-reading flowmeters feature a calibrated scale for a specific medium - no need to convert readings to get flow rate│Use with water for laboratory and industrial applications
Dual-scale flowmeters indicate gallons and liters simultaneously with an easy-to-read linear scale and highly visible red indicator.
$596.50 - $865.75USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (GPM) Process Connection (NPT(F))
Availability Pricing
COMPARE
0.5
$602.75USD / Each
COMPARE
1
$596.50USD / Each
COMPARE
2
$661.50USD / Each
COMPARE
5
$661.50USD / Each
COMPARE
15
$661.50USD / Each
COMPARE
10
$865.75USD / Each
COMPARE
10
$848.00USD / Each
COMPARE
20
$848.00USD / Each
Showing 1- 8 of 8