Masterflex® Direct Reading Variable-Area Flowmeters for Water, In-Line, Polysulfone Housing
Get better results than acrylic
  • Direct-reading flowmeters feature a calibrated scale for a specific medium - no need to convert readings to get flow rate│Use with water for laboratory and industrial applications
Choose from standard, high-capacity, or ultrapure in-line flowmeters. Standard models are designed for low flow readings—measure flow rates as low as 0.025 GPM. The high-capacity flowmeters measure flow rate as high as 130 GPM!
These large, durable flowmeters have 316 SS internal parts. New ultrapure flowmeters use only plastic wetted parts, making these great for ultrapure water such as deionized water. For high-capacity models with polycarbonate shields around the flowtube, call our Application Specialists and get more information.
$135.80 - $938.25USD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Fluid Type Max Flow Rate (GPM) Process Connection
Availability Pricing
COMPARE
Water 0.25 1/2" NPT(M)
$136.90USD / Each
COMPARE
Water 1 1/2" NPT(M)
$135.80USD / Each
COMPARE
Water 2 1/2" NPT(M)
$136.90USD / Each
COMPARE
Water 5 1/2" NPT(M)
$136.90USD / Each
COMPARE
Water 10 3/4" NPT(M)
$155.70USD / Each
COMPARE
Water 10 1" NPT(F)
$436.00USD / Each
COMPARE
Water 20 1" NPT(F)
$445.90USD / Each
COMPARE
Water 30 1" NPT(F)
$491.80USD / Each
COMPARE
Water 40 1" NPT(F)
$524.50USD / Each
COMPARE
Water 60 2" NPT(F)
$881.00USD / Each
COMPARE
Water 80 2" NPT(F)
$857.75USD / Each
COMPARE
Water 130 2" NPT(F)
$938.25USD / Each
Showing 1- 12 of 12