Masterflex® Direct Reading Variable Area Flowmeters for Water, PFA Housing
A unique combination of rugged frame and chemically inert wetted materials
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate│Use with water for laboratory and industrial applications
  • Rigid aluminum frame is PFA coated to resist ambient corrosives
  • Ideal for applications involving ultrapure water or highly corrosive solvents
These flowmeters are great for use with etchants and high purity water like those used in the semiconductor industry. Each flowmeter is individually tested on a mass spectrometer leak detector and certified to a leak integrity rating of 1x10–7 sccs helium or better. A polycarbonate front shield magnifies readings 16% for reading at greater distances. Scale measures 75-mm on flowmeters rated for 5,000 ml/min or less; 125-mm on flowmeters rated to 13 L/min and above. All models have direct-reading water scales.
$419.00 - $1,027.50USD / Each
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (mL/min) Min Flow Rate (mL/min) Pipe Size
Availability Pricing
COMPARE
75 5 1/4"
$419.00USD / Each
COMPARE
250 5 1/4"
$419.00USD / Each
COMPARE
400 50 1/4"
$419.00USD / Each
COMPARE
500 90 1/4"
$419.00USD / Each
COMPARE
1000 100 1/4"
$419.00USD / Each
COMPARE
2000 350 3/8"
$419.00USD / Each
COMPARE
2500 800 3/8"
$419.00USD / Each
COMPARE
3000 1250 3/8"
$419.00USD / Each
COMPARE
5000 1750 3/8"
$419.00USD / Each
COMPARE
75 5 1/4"
$585.50USD / Each
COMPARE
250 5 1/4"
$585.50USD / Each
COMPARE
400 50 1/4"
$585.50USD / Each
COMPARE
1000 100 1/4"
$585.50USD / Each
COMPARE
2000 350 3/8"
$585.50USD / Each
COMPARE
5000 1750 3/8"
$585.50USD / Each
COMPARE
13000 750 1/2"
$1,027.50USD / Each
COMPARE
$1,027.50USD / Each
Showing 1- 17 of 17