Masterflex® Direct-Reading, Variable-Area Flowmeters, Stainless Steel Housing
Ideal for the most aggressive process applications
  • Thick polycarbonate front shield provides protection
  • Unique design facilitates ease of maintenance
  • Easy-to-read direct-reading scale offers both metric and English measurements
  • Available in fluted or plain tapered tubes
Detachable, clear 3/16"-thick polycarbonate front shield provides protection at maximum rated temperature and pressure. Easily read the graduated direct-reading scale. The interchangeable transparent scale plates mount directly on the front of the flowtube. Unique float stops allow easy cleaning-remove and replace the float without disturbing either the flowtube or the shield. Tube sizes 3, 4, and 5 are fluted; tube sizes 6, 8, and 9 are tapered with a stainless steel guide rod.
$556.25 - $1,190.00USD / Each
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (GPM)
Availability Pricing
COMPARE
0.25
$556.25USD / Each
COMPARE
0.36
$556.25USD / Each
COMPARE
0.74
$556.25USD / Each
COMPARE
1
$556.25USD / Each
COMPARE
2
$556.25USD / Each
COMPARE
1.5
$556.25USD / Each
COMPARE
3.8
$644.50USD / Each
COMPARE
5
$644.50USD / Each
COMPARE
6
$644.50USD / Each
COMPARE
9.6
$644.50USD / Each
COMPARE
11
$644.50USD / Each
COMPARE
14
$644.50USD / Each
COMPARE
20
$644.50USD / Each
COMPARE
26
$644.50USD / Each
COMPARE
41
$1,190.00USD / Each
COMPARE
60
$1,190.00USD / Each
COMPARE
86
$1,190.00USD / Each
Showing 1- 17 of 17