Masterflex® Dual-Head Chemical Feed Solenoid Diaphragm Pumps
  • Flow rates from 37.9 to 669 mL/min
  • Overload-protected, fan-cooled, shaded-pole gear motor (1/35 hp) is permanently lubricated
  • Pumps feature two separate head assemblies—obtain proportional flow rates for two different materials or twice the maximum flow r
$913.00 - $943.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (mL/min) Max Flow Rate (Strokes/min) Max Pressure (PSI)
Availability Pricing
COMPARE
37.9 14 120
$943.00USD / Each
COMPARE
669 125 20
$913.00USD / Each
Showing 1- 2 of 2
Need Help with this product?
Call Us At1-847-549-7600
Live chat