Masterflex® End Suction Petroleum Centrifugal Pumps
Compatible with three distinct fuel classifications
  • Models for the following fluid groups:
    - Gasoline, kerosene, avgas, and jet fuel
    - Biodiesel, fuel oil, and diesel
    - Ethanol and E85 blend
  • Available in both self-priming and standard end suction configurations
  • Custom configurations are a phone call away to our Application Specialists
These close-coupled pumps have been specifically engineered for compatibility with clean, nonabrasive petroleum products. Transfer and delivery of various fuels such as gasoline, ethanol, biodiesel, and fuel oils are just a few of the petroleum-based products these pumps can handle.
$1,399.00 - $3,080.50USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (GPM) Max Head (ft) Motor hp
Availability Pricing
COMPARE
65 112 2
$3,080.50USD / Each
COMPARE
80 127 3
$3,076.50USD / Each
COMPARE
65 112 2
$1,399.00USD / Each
COMPARE
80 127 3
$1,511.00USD / Each
COMPARE
65 112 2
$3,035.00USD / Each
COMPARE
80 127 3
$3,077.00USD / Each