Masterflex® FDA Compliant Sanitary Drum Pumps
Capable of pumping fluids with viscosities up to 1000 cps
  • 316 SS construction with PTFE elastomers
  • Easy to clean with Tri-Clamp® fittings
The sanitary drum pumps are engineered to transfer low-viscosity food products, pharmaceutical materials, and personal care solutions from drums, totes, and kettles. Wetted surfaces are constructed with FDA-compliant materials which provide an ideal solution for sanitary applications.
$2,864.50 - $3,214.00USD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (GPM) Motor Type Description
Availability Pricing
COMPARE
32 ODP FDA-Compliant Sanitary Drum Pump, 32 GPM, 39"L SS Tube; ODP, 110V
$2,864.50USD / Each
COMPARE
32 ODP FDA-Compliant Sanitary Drum Pump, 32 GPM, 39"L SS Tube; ODP, 220V
$2,940.50USD / Each
COMPARE
32 TEFC FDA-Compliant Sanitary Drum Pump, 32 GPM, 39"L SS Tube; TEFC, 110V
$3,137.50USD / Each
COMPARE
32 TEFC FDA-Compliant Sanitary Drum Pump, 32 GPM, 39"L SS Tube; TEFC, 220V
$3,171.50USD / Each
COMPARE
26 Air FDA-Compliant Sanitary Drum Pump, 26 GPM, 39"L SS Tube; Air
$3,024.00USD / Each
COMPARE
15 ODP FDA-Compliant Sanitary Drum Pump, 15 GPM, 39"L SS Tube; ODP, 110V
$2,936.50USD / Each
COMPARE
15 TEFC FDA-Compliant Sanitary Drum Pump, 15 GPM, 39"L SS Tube; TEFC, 110V
$3,145.50USD / Each
COMPARE
15 TEFC FDA-Compliant Sanitary Drum Pump, 15 GPM, 39"L SS Tube; TEFC, 220V
$3,214.00USD / Each
COMPARE
12 Air FDA-Compliant Sanitary Drum Pump, 12 GPM, 39"L SS Tube; Air
$3,072.00USD / Each
Showing 1- 9 of 9