Masterflex® Fitting Assortment Packs
Bundled in assortment packs for convenience and to save you money!
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi). Just slip your flexible tubing or hose onto the barb to create a secure seal.
$64.00 - $205.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Description
Availability Pricing
COMPARE
Polypropylene T-connector assortment pack, PP, 6 each of sizes 4mm to 14 mm, 36 pieces total
$80.00USD / Pkg of 36
COMPARE
Polypropylene Y-connector assortment pack, PP, 6 each of sizes 4mm to 14 mm, 36 pieces total
$80.00USD / Pkg of 36
COMPARE
Polypropylene Hose Barb Fitting Kit, PP, Mini Assortment, 240 Pieces/Pk
$135.00USD / Pkg of 240
Scienceware 19570 Barbed fittings, Straight/reducing connector pack
Always in Stock
EW-06460-00 Mfr # 19570
COMPARE
Polypropylene Barbed fittings, Straight/reducing connector pack
$64.00USD / Pkg of 22
COMPARE
Polypropylene, Nylon, Polycarbonate Assorted luer fittings (477 pieces total)
$205.00USD / Each