Masterflex® Flow Sensor Flow Controllers
Easily control, view, and store your flow rates
  • Control your Low-Flow Impeller Sensors with these controllers
These new controllers display flow rate and total (resettable) simultaneously on two 8-digit Display LCDs. Push-button keys and menu prompts make it easy to program the controller. The 4-to-20 mA output lets you send data to a recorder or datalogger. Nonvolatile memory stores flow total in the event of power failure.
Select a 1¼4 DIN panel- or wall-mount controller depending on your application needs. Panel-mount model 33110-60 features a gasketed front panel, membrane switches, and NEMA Type 12 (IP52) enclosure. Wall-mount model 33110-70 has a clear cover and splashproof NEMA Type 4X (IP56) enclosure. Both models include a 21¼2-ft L cable with stripped ends. Order power supply for 110 VAC operation separately.
$595.00 - $654.75USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Output Power
Availability Pricing
COMPARE
12 to 24 VDC; K-factor: 0.050 to 1999.999 7-30Vdc, 4mA (4-20 mA whenloop-powered)
$595.00USD / Each
COMPARE
12 to 24 VDC; K-factor: 0.050 to 1999.999 7-30Vdc, 4mA (4-20 mA whenloop-powered)
$654.75USD / Each