Masterflex® Flowmeters and Controllers for Water
Achieve accurate, ultrafast volumetric measurement and control
  • Technology allows for installation without typical inlet/outlet straight-run requirements
  • Units provide data on flow and temperature
  • All functions are user-programmable through an integrated keypad
These meters measure flow via pressure drop across a laminar flow element (LFE). Because the flow element makes the flow stream laminar, placement in the process does not require straight pipe runs upstream or downstream of the meter, greatly simplifying installation. The LFEs also provide an outstanding turndown ratio of 50:1 giving the meter a very broad and accurate measuring range. The design provides an ultrafast response at start-up or input change—often within 100 milliseconds. The 0 to 5 VDC output allows transmission of the flow value to a remote display, recorder, or controller regulating a valve or pump.|For the flow controllers, an integrated PID controller directs the unit's response to process changes. Set the P and D parameters to tailor the unit's response. Flow setpoint is established with the optional set point control module, a 0 to 5 V signal, or an RS-232 input signal. For portable flow metering (not controlling) applications, order the battery pack separately. Units can be mounted via threaded taps in the meter body.|Meters and controllers feature dynamic display that simultaneously shows flow rate and fluid temperature, and (for controllers) the set point. For the units shown, both power and input/output signals are transmitted through a single multi-pin connector.
$1,507.50 - $2,364.50USD / Each
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (mL/min) Min Flow Rate (mL/min) Description
Availability Pricing
COMPARE
50 0 Flow controller, volumetric, for water, 0 to 50 mL/min
$2,271.00USD / Each
COMPARE
100 0 Flow controller, volumetric for water, 0 to 100 mL/min
$2,254.50USD / Each
COMPARE
200 0 Flow controller, volumetric, for water, 0 to 200 mL/min
$2,254.50USD / Each
COMPARE
500 0 Flow controller, volumetric, for water, 0 to 500 mL/min
$2,254.50USD / Each
COMPARE
1 0 Flowmeter, volumetric, for water, 0 to 1 mL/min
$2,254.50USD / Each
COMPARE
5 0 Flowmeter, volumetric, for water, 0 to 5 mL/min
$2,112.50USD / Each
COMPARE
10 0 Flowmeter, volumetric, for water, 0 to 10 mL/min
$1,801.50USD / Each
COMPARE
50 0 Flowmeter, volumetric, for water, 0 to 50 mL/min
$1,507.50USD / Each
COMPARE
100 0 Flowmeter, volumetric, for water, 0 to 100 mL/min
$1,581.00USD / Each
COMPARE
200 0 Flowmeter, volumetric, for water, 0 to 200 mL/min
$1,581.00USD / Each
COMPARE
500 0 Flowmeter, volumetric, for water, 0 to 500 mL/min
$1,571.00USD / Each
COMPARE
1000 0 Flowmeter, volumetric, for water, 0 to 1 L/min
$1,591.00USD / Each
COMPARE
5000 0 Flowmeter, volumetric, for water, 0 to 5 L/min
$1,995.00USD / Each
COMPARE
10000 0 Flowmeter, volumetric, for water, 0 to 10 L/min
$2,364.50USD / Each
Showing 1- 14 of 14