Masterflex® Gear Pump Systems with Calibration Column, Low Flow
Choose the pump system configured best for your needs
  • Maximum flow range from 0.0198 to 0.0436 GPM depending on model│Configured skid mounted on a steel base
This pulseless flow pump system can be configured in a variety of ways to accommodate flow rates from 0.0022 to 0.0436 GPM. Calibration Column ensures clear and precise confirmation of flow in your system. System operates on 115 VAC, 50/60 Hz.
$5,508.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (mL/min) Max Temperature (° F) Wetted Materials
Availability Pricing
COMPARE
74.9 350 316 stainless steel (SS) housing; carbon graphite gears; PTFE seals
$5,508.00USD / Each