Masterflex® Gear Pump Systems with Calibration Column, Relief Valve, and Static Mixer
Choose the pump system configured best for your needs
  • Maximum flow range from 0.5 to 10 GPM depending on model│Configured skid mounted on a steel base
This pulseless flow pump system can be configured in a variety of ways to accommodate flow rates from 0.1000 to 50.0 GPM. Calibration column ensures clear and precise confirmation of pump flow in your system. Relief valve protects your system from excessive pressure. Post dilute with static mixer has no moving parts. It allows water or another fluid to be mixed with the chemical being pumped at desired percentages — ideal for polymers and fluids with different density and viscosity parameters. System operates on 115 VAC, 50/60 Hz.
$12,411.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (mL/min) Max Temperature (° F) Wetted Materials
Availability Pricing
COMPARE
37850 600 single carbon/ceramic mechanical seal; 316 SS housing, main gear, and shafts; PTFE idler; PTFE wear plates and bearings
$12,411.00USD / Each