Masterflex® High-Precision, High-Pressure Piston Pumps
The right pump for your HPLC application
  • Flow rates from 10 to 300 mL/min Digital stepper motor ensures accuracy at low flow rates Remotely control pump with a PC via RS-232 and micro USB Self-flush feature washes piston with every stroke Interactive keypad controls make programming at the pump
Single-Head Pumps precisely meter and dispense chemicals, perfect for small-scale laboratory fluid transfers and HPLC applications. Small footprint saves bench and fume hood space.
High-Flow Dual-Head Pumps are ideally suited to Preparative
$3,303.50 - $10,480.00USD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (mL/min) Max Pressure (PSI) Wetted Materials
Availability Pricing
COMPARE
10 2000 Synthetic ruby, sapphire, UHMWPE, PTFE
$3,340.00USD / Each
COMPARE
10 2000 Synthetic ruby, sapphire, UHMWPE, PTFE
$3,303.50USD / Each
COMPARE
100 4000 Synthetic ruby, sapphire, UHMWPE, PTFE
$10,090.00USD / Each
COMPARE
100 4000 Synthetic ruby, sapphire, UHMWPE, PTFE
$9,949.00USD / Each
COMPARE
300 1000 Synthetic ruby, sapphire, UHMWPE, PTFE
$10,480.00USD / Each
COMPARE
10 6000 Synthetic ruby, sapphire, UHMWPE, PTFE
$6,101.00USD / Each
COMPARE
100 4000 Synthetic ruby, sapphire, UHMWPE, PTFE
$9,013.00USD / Each
COMPARE
12 6000 Synthetic ruby, sapphire, UHMWPE, PTFE
$8,729.00USD / Each
COMPARE
10 5000 Synthetic ruby, sapphire, UHMWPE, PTFE
$5,195.00USD / Each
Showing 1- 9 of 9