Masterflex® High-Purity Plastic Diaphragm Valves
Fully drainable and easy to maintain!
  • Eliminate leaks—ideal for food or pharmaceutical processing
  • Compatible with Tri-Clamp® connections for easy maintenance, change out, and orientation—no cutting or welding needed!
  • Choose either natural polypropylene (PP) or Kynar® (PVDF) body—both are excellent for ultrapure applications
  • Models with Kynar® (PVDF) body offer additional chemical resistance to corrosives
These high-purity valves feature manual actuators. Maximum viscosity is 400 cSt. Maximum fluid temperature is 180°F (82°C).
$704.00 - $1,572.00USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Clamp Size (in) Cv Wetted Materials
Availability Pricing
COMPARE
0.75 4.0 Polypropylene, EPDM
$704.00USD / Each
COMPARE
1 12.3 Polypropylene, EPDM
$844.00USD / Each
COMPARE
1.5 21.0 Polypropylene, EPDM
$1,068.00USD / Each
COMPARE
2 29.2 Polypropylene, EPDM
$1,175.00USD / Each
COMPARE
0.75 4.0 PVDF, EPDM
$949.00USD / Each
COMPARE
1 12.3 PVDF, EPDM
$1,560.00USD / Each
COMPARE
1.5 21.0 PVDF, EPDM
$1,572.00USD / Each
Showing 1- 7 of 7