Masterflex® High-Viscosity Sanitary Drum Pumps
Capable of pumping fluids with viscosities up to 10,000 cps
  • Progressive cavity design allows for low-shear properties
  • Flow rates up to 12 GPM (45 LPM)
  • 316 stainless steel construction with PTFE elastomers
These drum pumps are engineered specifically for transferring food products, pharmaceuticals, and cosmetic products from barrels, kettles, and tanks. They offer a progressive cavity, positive-displacement delivery system resulting in a continuous flow of material with little product degradation. This unique design allows you to quickly dismantle, clean, and re-assemble the pump.
$7,306.00 - $10,365.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (GPM) Max Viscosity (cps) Tube Length, Inlet (in)
Availability Pricing
COMPARE
12 10000 39
$7,306.00USD / Each
COMPARE
6 10000 39
$10,365.00USD / Each
COMPARE
4 10000 39
$7,859.00USD / Each