Masterflex® I/P® Digital Process Pumps
Precision digital drives offer three dispensing modes and precision low-maintenance motors
  • Flow range is 0.001 LPM to 8.0 LPM with included tubing
  • Housings are NEMA 4X (IP66) rated for washdown
  • Graphical LCD shows four operating modes continuous run, timed dispense, copy dispense, volume dispense. View pump performance continually.
  • Menu-driven interface lets you select one of seven languages for programming and operation
  • Dispense by Volume in mL (0.001 to 99999 mL), L, or gal; Copy—1 to 9,999 dispense cycles; or Time—time interval of on/off cycles (1 sec to 99 hr, 59 min, 59 sec per cycle)
  • Reversible motor—purge tubing before or after pumping; pump fluid in either direction
  • Brushless, maintenance-free motor offers ±0.1% speed control accuracy with a 6000 to 1 turndown ratio
  • Remote control capabilities via watertight connection
  • – Speed control input scalable 0 to 20 mA, 4 to 20 mA, or 0 to 10 V
  • – Pumping direction requires open collector or contact closure
  • – Start/stop/purge requires open collector or contact closure
  • – Tachometer output scalable 0 to 20 mA, 4 to 20 mA, 0 to 10 V, open collector
These pump systems give you rugged performance and precise digital control. Ideal for fluid transfer, dispensing, and metering applications in laboratory, process, and industrial environments including pharmaceutical, food & beverage, dairy, chemical, manufacturing, and water treatment. The NEMA 4X (IP66) rated housings are completely sealed, easy-to-clean, and resistant to commonly used cleaning and sanitizing solutions.
Precision drives feature a programmable dispense interval that lets you set the delay between dispense cycles for convenient, automated dispensing by time or by volume. Programmed calibration ensures dispense and display accuracy—drive stores one calibration value per tubing size. Program the drive via the simple membrane keypad; keypad lock/unlock feature protects settings. Select tubing size and enter desired flow rate—drive locks in required rpm.
$5,965.00 - $6,790.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Housing Material Max Flow Rate Per Channel (LPM) Open-Head Sensor
Availability Pricing
COMPARE
Stainless Steel 8 No
$6,455.00USD / Each
COMPARE
Stainless Steel 8 Yes
$6,790.00USD / Each
COMPARE
Powder-Coat Steel 8 No
$5,965.00USD / Each
COMPARE
Powder-Coat Steel 8 Yes
$6,265.00USD / Each