Masterflex® I/P® Fixed-Speed Wash-Down Drives
Easy operation and easy
on the budget
  • Purge system or pump in either direction with combination on/off/reverse switch
  • Constructed of painted steel/aluminum IP55-rated housing
  • Get more flow—capable of driving multiple pump heads for multichannel pumping
These fixed-speed drives are IP55-rated for protection from dust and water. Accept I/P standard, Easy-Load®, or High-Performance pump heads. Stack up to two Standard or Easy-Load pump heads to double flow.
$2,745.00 - $2,925.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (LPM) Max RPM (rpm) Power (VAC)
Availability Pricing
COMPARE
14.1 540 115
$2,745.00USD / Each
COMPARE
11.8 450 230
$2,925.00USD / Each
COMPARE
2.6 100 115
$2,760.00USD / Each