Masterflex® I/P® High-Performance Precision Pump Tubing, Tygon® S3™ E-LFL
Long pumping life with superior flexibility reduces downtime and tubing failure
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory Approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • Extremely low spallation for reduced fluid contamination
Masterflex® Tygon® E-FLF High-Performance Precision pump tubing is a non-DEHP formulation specially formulated for even longer pumping life with superior flexibility. This tubing features low particulate spallation and no phthalates or BPA, protecting fluid purity. Ideal for food and beverage, cosmetics, and pharmaceutical applications, this tubing complies with FDA, USP Class VI, and EU Foods standards.
$531.00 - $696.00USD / Pkg of 1
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Hose Barb Size (in) I/P Masterflex tubing size Max Flow Rate (mL/min)
Availability Pricing
COMPARE
3/8 70 8000
$531.00USD / Pkg of 1
COMPARE
1/2 88 17000
$665.00USD / Pkg of 1
COMPARE
5/8 89 19000
$696.00USD / Pkg of 1