Masterflex® I/P® Long-Shaft Pump Head for Precision Tubing
Ideal for fluid transfer applications requiring high accuracy and repeatability
  • Designed and optimized for one tubing size
  • Three roller design for smooth flow
  • Choose from stainless steel (SS) or cold-rolled steel (CRS) rotor / roller for greater durability and chemical resistance
  • Pump heads are stackable for multichannel applications
  • Compact design for efficient use of space
Standard heads are precision molded to accommodate a single size of tubing—use for common fluid transfer applications requiring accuracy and excellent repeatability. Available with clear polycarbonate (PC) housing with either cold-rolled steel (CRS) or stainless steel (SS) rotors; CRS offers economy, while SS offers better chemical resistance and durability.
Use with Masterflex® I/P® pumps requiring the long-shaft style pump head. These pump heads are not compatible with standard shaft drives.
Pump head is compact and simple in design with same side tubing entry/exit—perfect for space-limited applications. Standard heads are compact and easy to integrate into your OEM applications. Call for more information on our comprehensive OEM services.
$595.00 - $1,195.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (LPM) Rotor Material Tubing sizes accepted
Availability Pricing
COMPARE
8 Cold-rolled Steel I/P® 73
$595.00USD / Each
COMPARE
13 Cold-rolled Steel I/P® 82
$625.00USD / Each
COMPARE
13 Stainless Steel I/P® 82
$870.00USD / Each
COMPARE
13 Stainless Steel I/P® 26, I/P 73, I/P 82
$1,195.00USD / Each
COMPARE
13 Cold-rolled Steel I/P® 26, I/P 73, I/P 82
$895.00USD / Each
COMPARE
13 Stainless Steel I/P® 26, I/P 73, I/P 82
$1,105.00USD / Each