Masterflex® I/P® Precision Pump Tubing, Norprene®
Outlast and outperform other tubing with 10,000 hours of tubing life
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory Approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • Opaque beige color protects light and UV sensitive samples
Masterflex® Norprene® Precision pump tubing is specially formulated not to weaken or crack after years of exposure to heat and ozone, providing longer peristaltic pump life. This tubing is heat sealable and weldable and able to withstand a broad range of temperature applications. Able to outperform other general-purpose tubing, Norprene features superior abrasion and flexural fatigue resistance, making it a better alternative to rubber tubing.
$187.00 - $291.00USD / Pkg of 1
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Hose Barb Size (in) I/P Masterflex tubing size Max Flow Rate (mL/min)
Availability Pricing
COMPARE
1/4 26 4000
$187.00USD / Pkg of 1
COMPARE
3/8 73 8000
$220.00USD / Pkg of 1
COMPARE
1/2 82 13000
$291.00USD / Pkg of 1