This product is now discontinued, View Related Products
Masterflex® I/P® Precision Pump Tubing, PharMed® BPT, Double-Y Elements
Protect your light-sensitive samples
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Applications: Cell and tissue culture work, pharmaceutical, light-sensitive samples
Characteristics: Nontoxic, nonhemolytic, extremely low permeability, resists acids, bases, and oxidizing agents; heat-sealable, bondable, formable, opaque beige
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Hose Barb Size (in) I/P Masterflex tubing size Description
Availability Pricing
COMPARE
1/4 26 I/P® Precision Pump Tubing, PharMed® BPT Plus PVDF Fitting, Double Y, I/P 26
Call For Price
COMPARE
3/8 73 I/P® Precision Pump Tubing, PharMed® BPT Plus PVDF Fitting, Double Y, I/P 73
Call For Price
COMPARE
1/2 82 I/P® Precision Pump Tubing, PharMed® BPT Plus PVDF Fitting, Double Y, I/P 82