Masterflex® I/P® Sanitary Fitting Precision Pump Tubing Assembly, PharMed® BPT
Protect your light-sensitive samples
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Applications: Cell and tissue culture work, pharmaceutical, light-sensitive samples
Characteristics: Nontoxic, nonhemolytic, extremely low permeability, resists acids, bases, and oxidizing agents; heat-sealable, bondable, formable, opaque beige
Requires sanitary gaskets for both connections to ensure quality seal
$179.00 - $277.00USD / Pkg
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
I/P Masterflex tubing size Tubing ID (mm)
Availability Pricing
COMPARE
26 6.4
$241.00USD / Pkg
COMPARE
70 9.5
$248.00USD / Pkg
COMPARE
73 9.5
$203.00USD / Pkg
COMPARE
82 12.7
$203.00USD / Pkg
COMPARE
88 12.7
$184.00USD / Pkg
COMPARE
89 15.88
$179.00USD / Pkg
COMPARE
26 6.4
$263.00USD / Pkg
COMPARE
70 9.5
$276.00USD / Pkg
COMPARE
73 9.5
$237.00USD / Pkg
COMPARE
82 12.7
$263.00USD / Pkg
COMPARE
88 12.7
$277.00USD / Pkg
COMPARE
73 9.5
$217.00USD / Pkg of 1
Showing 1- 12 of 12