Masterflex® I/P® Spooled Pump Tubing, BioPharm Platinum-Cured Silicone
Smooth inner surface designed to reduce risk of particle entrapment and contamination
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory Approvals to match your unique application
Choose our BioPharm Silicone platinum-cured tubing for purity, very low extractables, and an ultra-smooth inner surface. Ideal for pharmaceutical and biotechnological applications, this tubing complies with USP class VI and 3A sanitary standards.
The ultra-smooth inner surface minimizes particle entrapment and microscopic build-up during fluid transfer. Tubing can be sterilized by autoclave, gamma radiation, or ethylene oxide.
Spools contain continuous pump tubing, allowing you to cut the exact length needed and eliminating waste.
$1,543.00 - $5,243.00USD / Pkg of 1
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Hose Barb Size (in) I/P Masterflex tubing size Max Flow Rate (mL/min)
Availability Pricing
COMPARE
1/4 26 4000
$5,243.00USD / Pkg of 1
COMPARE
3/8 73 8000
$1,543.00USD / Pkg of 1
COMPARE
1/2 88 17000
$3,221.00USD / Pkg of 1
COMPARE
5/8 89 19000
$4,534.00USD / Pkg of 1