Masterflex® In-Line Direct-Reading, Variable-Area Flowmeters for Water, Acrylic Housing, Dual Scales
Read measurements in an unobstructed 360° view
  • Easy-to-read dual scales (GPM/LPM)
  • Extremely durable—machined from high-quality acrylic rod stock
  • One-piece, in-line body occupies less space for a clean installation
These meters are designed for use in more aggressive process environments. In addition to the machined acrylic bodies, the design utilizes stainless steel float guides. The combination is a meter suitable for high-vibration, high-fluctuation process applications. Some versions include polypropylene end fittings reinforced with aluminum stress rings for added strength to the meters’ design.
$135.10 - $357.60USD / Each
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
Compareitem specifications
Fluid Type Max Flow Rate (GPM) Process Connection
Availability Pricing
COMPARE
Water 0.25 1/4" NPT(F)
$158.60USD / Each
COMPARE
Water 0.25 3/8" NPT(F)
$155.40USD / Each
COMPARE
Water 1 3/8" NPT(F)
$139.10USD / Each
COMPARE
Water 1 1/2" NPT(F)
$137.80USD / Each
COMPARE
Water 2 3/8" NPT(F)
$135.10USD / Each
COMPARE
Water 2 1/2" NPT(F)
$139.10USD / Each
COMPARE
Water 3 3/8" NPT(F)
$146.80USD / Each
COMPARE
Water 3 1/2" NPT(F)
$151.30USD / Each
COMPARE
Water 5 3/8" NPT(F)
$139.10USD / Each
COMPARE
Water 5 1/2" NPT(F)
$136.90USD / Each
COMPARE
Water 10 3/4" NPT(F)
$259.40USD / Each
COMPARE
Water 10 1" NPT(F)
$256.80USD / Each
COMPARE
Water 20 3/4" NPT(F)
$259.40USD / Each
COMPARE
Water 20 1" NPT(F)
$251.80USD / Each
COMPARE
Water 25 1-1/2" NPT(M)
$357.60USD / Each
COMPARE
Water 40 1-1/2" NPT(M)
$357.60USD / Each
COMPARE
Water 50 1-1/2" NPT(M)
$357.60USD / Each
Showing 1- 17 of 17