Masterflex® Infusion Syringe Pumps, High-Pressure
Ideal for low-flow, higher pressure applications where precision is needed
  • Flow rates up to 145.5 mL/min
  • RS-232 connectivity for interface to a remote computer
  • Greater than 100 lbs of linear force
These syringe pumps more than double the linear force available on other versions. This extra force makes the pump ideal for fluid delivery to reactors in chemical applications or for working with viscous fluids. The robust design of the syringe holder en
$4,955.00 - $5,497.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate Per Syringe (mL/min) Min Flow Rate Per Syringe (microL/min) Number of Syringes
Availability Pricing
COMPARE
145.5
$4,955.00USD / Each
COMPARE
145.5
$5,497.00USD / Each
COMPARE
145.5
$5,105.00USD / Each