Masterflex® L/S® Analog Variable-Speed Console Drive Systems
Compact, cost effective pump systems for general fluid transfer applications
  • Simple operation with single-turn speed control
  • Separate power switch enables you to maintain speed setting when turning drive on/off; front-panel LED indicates power on
  • Reversible motor lets you easily prime/purge tubing and pump in either direction
  • Remote control of start/stop via DB9 connector on back of drive
Choose a convenient, complete system all with one catalog number.
$1,498.00 - $1,578.00USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate Per Channel (mL/min) Min Flow Rate Per Channel (mL/min) Power (VAC)
Availability Pricing
COMPARE
480 16 115
$1,574.00USD / Each
COMPARE
480 16 230
$1,574.00USD / Each
COMPARE
1700 56 115
$1,578.00USD / Each
COMPARE
1700 56 230
$1,578.00USD / Each
COMPARE
480 16 115
$1,498.00USD / Each
COMPARE
480 16 230
$1,498.00USD / Each
COMPARE
1700 56 115
$1,502.00USD / Each
COMPARE
1700 56 230
$1,502.00USD / Each
Showing 1- 8 of 8