Masterflex® L/S ® Bulk-Packed Pump Tubing, Platinum-Cured Silicone
Excellent biocompatibility that meets multiple classifications
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Slightly greater clarity than peroxide-cured tubing
  • Smooth surfaces; lower protein binding levels
  • Fewer potential leachables
Applications: Pharmaceutical, biotech, peristaltic pumping, and food applications
Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic, translucent
Our bulk tubing comes in convenient cases containing up to 20 sealed bags of tubing (depending on tubing size). Bulk tubing eliminates individual cardboard boxes, reduces unpacking time, and saves you money on shipping. Tubing comes in 25-ft bags.
$1,305.00 - $2,528.00USD / Case of 1
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Hose Barb Size (in) L/S Masterflex tubing size Max Flow Rate (mL/min)
Availability Pricing
COMPARE
1/16 13 36
$1,426.00USD / Case of 1
COMPARE
1/16 14 130
$1,671.00USD / Case of 1
COMPARE
3/16 15 1100
$2,166.00USD / Case of 1
COMPARE
1/8 16 480
$1,605.00USD / Case of 1
COMPARE
1/4 17 1700
$2,094.00USD / Case of 1
COMPARE
3/8 18 2300
$1,305.00USD / Case of 1
COMPARE
1/4 24 1800
$2,528.00USD / Case of 1
COMPARE
3/16 25 1000
$2,092.00USD / Case of 1
COMPARE
3/8 35 2600
$1,686.00USD / Case of 1
COMPARE
3/8 36 3400
$2,029.00USD / Case of 1
Showing 1- 10 of 10