Masterflex® L/S® Easy-Load® Pump Heads for High-Performance Precision Tubing
Save time with fast tubing changes and reduced maintenance
  • New design and automatic tubing retention allow for multiple tubing sizes—change tubing without removing head from drive
  • Rotor is constructed of stainless steel (SS) for enhanced durability
  • Pump heads are stackable for multichannel applications
  • Compact design for efficient use of space
  • Suitable for use with L/S® High-Performance Precision pump tubing sizes
This Easy-Load® pump head accepts high-performance precision tubing— ensures optimal performance from your Masterflex pump. Select a pump head for one of the following precision tubing sizes: L/S 15 or L/S 24.
$428.00 - $518.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (mL/min) Open-Head Sensor Tubing sizes accepted
Availability Pricing
COMPARE
1700 No L/S® 15, L/S 24
$428.00USD / Each
COMPARE
1700 Yes L/S® 15, L/S 24
$518.00USD / Each