Masterflex® L/S® High-Performance Precision BioPharm Silicone Platinum-Cured Pump Tubing

Formulation: Tygon 3350 silicone, platinum cured.
Applications: Pharmaceutical, cosmetic, research, and auto-analysis applications.
Characteristics: Ultra-smooth; minimizes bacterial growth. Good for mild to medium concentration bases, salts, and alcohols; odorless, tasteless, and nontoxic. Transparent.
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Hose Barb Size (in) L/S Masterflex tubing size Max Flow Rate (mL/min)
Availability Pricing
COMPARE
3/16 15 1100
$202.00USD / Pkg of 1
COMPARE
1/4 24 1800
$230.00USD / Pkg of 1
COMPARE
3/8 35 2600
$248.00USD / Pkg of 1
COMPARE
3/8 36 3400
$287.00USD / Pkg of 1
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat