Masterflex® L/S® High-Performance Precision Pump Tubing, C-Flex® ULTRA
Longest life of any C-Flex® formulation
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
Applications: General-purpose lab uses requiring high performance and long life
Characteristics: Exceptional tensile and tear strength, good biocompatibility, low spallation, reduced residue and low gas permeability, smooth surface, translucent
$52.00 - $154.00USD / Pkg of 1
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Hose Barb Size (in) L/S Masterflex tubing size Max Flow Rate (mL/min)
Availability Pricing
COMPARE
3/16 15 1100
$103.00USD / Pkg of 1
COMPARE
1/4 24 1800
$115.00USD / Pkg of 1
COMPARE
3/8 35 2600
$152.00USD / Pkg of 1
COMPARE
3/8 36 3400
$154.00USD / Pkg of 1
COMPARE
1/4 24 1800
$52.00USD / Pkg of 1