This product is now discontinued, View Related Products
Masterflex® L/S® High-Performance Precision Pump Tubing, PharMed® BPT, Double-Y Elements
Protect your light-sensitive samples
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Applications: Cell and tissue culture work, pharmaceutical, light-sensitive samples
Characteristics: Nontoxic, nonhemolytic, extremely low permeability, resists acids, bases, and oxidizing agents; heat-sealable, bondable, formable, opaque beige
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Hose Barb Size (in) L/S Masterflex tubing size Description
Availability Pricing
COMPARE
3/16 15 L/S® High-Performance Precision Pump Tubing, PharMed® BPT and PVDF Fitting, Double Y, L/S 15
Call For Price
COMPARE
1/4 24 L/S® High-Performance Precision Pump Tubing, PharMed® BPT and PVDF Fitting, Double Y, L/S 24
Call For Price
COMPARE
3/8 35 L/S® High-Performance Precision Pump Tubing, PharMed® BPT and PVDF Fitting, Double Y, L/S 35
Call For Price
COMPARE
3/8 36 L/S® High-Performance Precision Pump Tubing, PharMed® BPT and PVDF Fitting, Double Y, L/S 36