Masterflex® L/S® MiniFlex® OEM Pump Heads
Pump head accepts continuous lengths of tubing for clean flow paths
  • Flow range of 1.8 to 220 mL/min—flow rate depends on drove rpm, pump head, and tubing size
  • Pump head accepts continuous lengths of tubing for clean flow paths
  • Dual-channel model delivers synchronous flow from two separate channels; single-channel model delivers highest flow rate
  • Automatic tubing retention makes tube loading quick and easy
The Masterflex® Miniflex® pump heads feature a wide occlusion angle that prevents fluid backflow up to the rated pressure of the tubing. Self-lubricating thermoplastic polyester rollers are low friction and long wearing for smooth operation and long life. A compact overall size envelope makes these heads ideal for OEM applications and integration into fluid systems and machinery.
$236.00 - $252.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate Per Channel (mL/min) Number of Channels Tubing sizes accepted
Availability Pricing
COMPARE
315 1 L/S® 13, L/S 14, L/S 16, L/S 25
$236.00USD / Each
COMPARE
162 2 L/S® 13, L/S 14, L/S 16
$252.00USD / Each