Masterflex® L/S® Multichannel Pump Head Cartridges
Use with L/S® multichannel cartridge pump heads for synchronous, multichannel pumping
  • Only Silicone and C-Flex® tubing formulations are recommended for size L/S® 17 tubing when used in L/S® cartridge pump heads.
Cartridges feature accurate flow rates and adjustable tubing retainers to hold tubing in place — for large cartridges, set tubing retainers to your tube size and you are ready to pump. On small cartridges, optimum retention is preset — simply load tubing and start pumping.
Cartridges are constructed from polycarbonate and glass-filled nylon. Mix small and large cartridges on pump heads that accept both sizes; two small cartridges occupy the same space as one large cartridge.
$122.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Tubing sizes accepted
Availability Pricing
COMPARE
L/S® 14, L/S 16, L/S 25, L/S 17
$122.00USD / Each
COMPARE
L/S® 14, L/S 16, L/S 25, L/S 17
$122.00USD / Each
COMPARE
Microbore two-stop tubing sets, L/S 13, L/S 14
$122.00USD / Each
COMPARE
Microbore two-stop tubing sets
$122.00USD / Each