Masterflex® L/S® Precision PharmaPure Pump Tubing
Ultra low spallation protects fluid purity – ideal for sensitive and costly fluids and for filtration applications
  • Nontoxic and nonhemolytic with excellent biocompatibility
  • Long pumping life; pump at continuous pressures up to 40 psi
  • Low extractables and low gas permeability
  • Meets USP Class VI criteria
Applications: Pharmaceutical and biotech
Characteristics: Long life under pressure up to 60 psi (4 bar), excellent biocompatibility, low extractables, very low spallation. Opaque off-white
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Hose Barb Size (in) L/S Masterflex tubing size Max Flow Rate (mL/min)
Availability Pricing
COMPARE
1/16 13 36
$128.00USD / Pkg of 1
COMPARE
1/16 14 130
$341.00USD / Pkg of 1
COMPARE
1/8 16 480
$136.00USD / Pkg of 1
COMPARE
1/4 17 1700
$211.00USD / Pkg of 1
COMPARE
3/8 18 2300
$228.00USD / Pkg of 1
COMPARE
3/16 25 1000
$231.00USD / Pkg of 1