Masterflex® L/S® Precision Pump Tubing, C-Flex®
Combines biocompatibility of silicone with chemical resistance similar to Tygon®
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Very low protein binding
  • Heat sealable, weldable
  • Economical tubing
Applications: General-purpose lab uses requiring high performance and long life
Characteristics: Exceptional tensile and tear strength, good biocompatibility, low gas permeability, smooth surface, opaque white
$53.00 - $92.00USD / Pkg of 1
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Hose Barb Size (in) L/S Masterflex tubing size Max Flow Rate (mL/min)
Availability Pricing
COMPARE
1/16 13 36
$53.00USD / Pkg of 1
COMPARE
1/16 14 130
$57.00USD / Pkg of 1
COMPARE
1/8 16 480
$62.50USD / Pkg of 1
COMPARE
1/4 17 1700
$84.50USD / Pkg of 1
COMPARE
3/8 18 2300
$92.00USD / Pkg of 1
COMPARE
3/16 25 1000
$80.50USD / Pkg of 1