Masterflex® L/S® Precision Washdown Modular Drives with Wall-Mount Controller
Place where convenient—ideal for limited spaces, hoods, or isolation chambers
  • Flow rate 0.06 to 3400 mL/min with L/S tubing—flow rate depends on drive rpm and tubing size
  • Three digit LED display shows motor rpm — confirm speed setting at a glance
  • Reversible motor—purge tubing before or after pumping; pump fluid in either direction
  • LED indicators confirm motor direction
  • Wall-mount controller housing is powder-coated aluminum, IP66/NEMA 4X rated for washdown
  • Remote control capabilities via weather-resistant connection on controller
L/S® modular drives feature reversible pumping and allow you the flexibility to separate the motor from the controller. These accurate modular drives have powerful motors, speed display, and ±0.25% speed control for precise flow regulation—ideal for metering applications.
$2,415.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (mL/min) Max RPM (rpm) No. of Pump Heads Accepted
Availability Pricing
COMPARE
3400 600 2
$2,415.00USD / Each
COMPARE
580 100 4
$2,415.00USD / Each