Masterflex® L/S® Replacement Brushes and Brush Caps
Quickly and easily replace components of your Masterflex system
  • Choose replacement brushes and caps based on your Masterflex L/S drive
  • Brushes should be inspected every six months or 2000 hours—whichever comes first
Easy to install, these replacement drive parts will help extend the life of your drive.
$36.00 - $86.00USD / Set of 1
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Drive Replacement Brush Cap
$36.00USD / Set of 1
COMPARE
Brushes for Peristaltic Pump Drives
$50.00USD / Set of 1
COMPARE
Replacement brushes for L/S drive series: 07554-80, -85, -90, -95
$86.00USD / Pkg of 2
COMPARE
L/S® Replacement Brushes, 2/PK
$60.50USD / Pkg of 2