Masterflex® L/S® Spooled Pump Tubing, BioPharm Platinum-Cured Silicone
Ultra-smooth inner surface minimizes particle entrapment
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
Applications: Pharmaceutical, cosmetic, research, and auto-analysis applications
Characteristics: Ultra-smooth, minimizes bacterial growth, good for mild to medium concentration bases, salts, and alcohols; odorless, tasteless, nontoxic, transparent
Spooled pump tubing provides continuous lengths of pump tubing on convenient, manageable spools that are easy to transport and require less storage space than the equivalent amount of individual tubing coils.
$1,944.00 - $3,803.00USD / Pkg of 1
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Hose Barb Size (in) L/S Masterflex tubing size Max Flow Rate (mL/min)
Availability Pricing
COMPARE
1/16 14 130
$1,944.00USD / Pkg of 1
COMPARE
3/16 15 1100
$3,303.00USD / Pkg of 1
COMPARE
1/8 16 480
$2,075.00USD / Pkg of 1
COMPARE
1/4 17 1700
$3,022.00USD / Pkg of 1
COMPARE
3/8 18 2300
$3,189.00USD / Pkg of 1
COMPARE
1/4 24 1800
$3,803.00USD / Pkg of 1
COMPARE
3/16 25 1000
$2,591.00USD / Pkg of 1
Showing 1- 7 of 7