Masterflex® L/S® Spooled Pump Tubing, PharMed® BPT
Protect your light-sensitive samples
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
Applications: Cell and tissue culture work, pharmaceutical, light-sensitive samples
Characteristics: Nontoxic, nonhemolytic, extremely low permeability, resists acids, bases, and oxidizing agents; heat-sealable, bondable, formable, opaque beige
Spooled pump tubing provides continuous lengths of pump tubing on convenient, manageable spools that are easy to transport and require less storage space than the equivalent amount of individual tubing coils.
$876.00 - $2,858.00USD / Pkg of 1
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Hose Barb Size (in) L/S Masterflex tubing size Max Flow Rate (mL/min)
Availability Pricing
COMPARE
1/16 14 130
$2,241.00USD / Pkg of 1
COMPARE
3/16 15 1100
$1,991.00USD / Pkg of 1
COMPARE
1/8 16 480
$2,324.00USD / Pkg of 1
COMPARE
1/4 17 1700
$2,858.00USD / Pkg of 1
COMPARE
3/8 18 2300
$2,556.00USD / Pkg of 1
COMPARE
1/4 24 1800
$2,031.00USD / Pkg of 1
COMPARE
3/8 35 2600
$2,209.00USD / Pkg of 1
COMPARE
3/8 36 3400
$876.00USD / Pkg of 1
Showing 1- 8 of 8