Masterflex® L/S® Variable-Speed Economy Modular Drive Systems
Versatile variable-speed pump system with a wide flow range and simple controls
  • Separate motor and controller are connected via a 6-ft (1.9 m) cable — place components where convenient or where space allows
  • Single-turn speed control and on/off switch with green power indicator — maintain speed setting while turning pump on/off
  • The 1/10-hp reversible motor has soft start and back EMF for ±2% speed control
  • Painted steel housing is IP33 rated for protection from vertically falling water
Choose a convenient, complete system all with one catalog number. A perfect upgrade from a fixed-speed drive, these variable-speed drives offer a wider rpm range and ±2% speed regulation.
$1,694.00 - $1,770.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate Per Channel (mL/min) Min Flow Rate Per Channel (mL/min) Pump Head
Availability Pricing
COMPARE
480 4.8 77800-60 (L/S® Easy-Load 3 Pump Head)
$1,694.00USD / Each
COMPARE
480 4.8 77800-60 (L/S® Easy-Load 3 Pump Head)
$1,704.00USD / Each
COMPARE
1700 56 77200-62 (L/S® Easy-Load II Pump Head)
$1,760.00USD / Each
COMPARE
1700 56 77200-62 (L/S® Easy-Load II Pump Head)
$1,770.00USD / Each